Product Details

SNP ID
rs7596744
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:151808244 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGATATAAAATTAAGGTTTTGGTGC[A/C]ACACTTGCAAGTTTTCACAACTGTT
Phenotype
MIM: 608960
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
ARL5A PubMed Links

Gene Details

Gene
ARL5A
Gene Name
ADP ribosylation factor like GTPase 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037174.1 Intron NP_001032251.1
NM_012097.3 Intron NP_036229.1
NM_177985.2 Intron NP_817114.2

View Full Product Details