Product Details

SNP ID
rs9261132
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30013286 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTCCACAAGATTCTCTGTCCCAAA[C/T]TGTTAATAGTGCTGCTGTTGAGAAA
Phenotype
MIM: 615801 MIM: 615714
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
HCG8 PubMed Links
Additional Information
For this assay, SNP(s) [rs74295253] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
HCG8
Gene Name
HLA complex group 8
There are no transcripts associated with this gene.

Gene
HLA-J
Gene Name
major histocompatibility complex, class I, J (pseudogene)
There are no transcripts associated with this gene.

Gene
ZNRD1ASP
Gene Name
zinc ribbon domain containing 1 antisense, pseudogene
There are no transcripts associated with this gene.

View Full Product Details