Product Details

SNP ID
rs9353486
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:87398767 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAGGAAACTGACAGTTCAGCCCTC[C/G]TCAACTGAGAGACAGTTGAATTAAA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CFAP206 PubMed Links

Gene Details

Gene
CFAP206
Gene Name
cilia and flagella associated protein 206
There are no transcripts associated with this gene.

Gene
LINC01590
Gene Name
long intergenic non-protein coding RNA 1590
There are no transcripts associated with this gene.

View Full Product Details