Product Details
- SNP ID
-
rs9658649
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:92926757 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTGCTGGGCTGGGAGGCTAGGACA[C/T]GGGTGTGTGGCTTTTGGTGGAATTA
- Phenotype
-
MIM: 118910
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CHGA
PubMed Links
Gene Details
- Gene
- CHGA
- Gene Name
- chromogranin A
View Full Product Details