Product Details

SNP ID
rs5986968
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:154302012 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCCTGAAACTGTCAGGATCATGTT[C/G]GTTTCCTCGCTGTTTGCCGGCTATT
Phenotype
MIM: 300092 MIM: 300044
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TEX28 PubMed Links

Gene Details

Gene
TEX28
Gene Name
testis expressed 28
There are no transcripts associated with this gene.

Gene
TKTL1
Gene Name
transketolase like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145933.1 Intron NP_001139405.1
NM_001145934.1 Intron NP_001139406.1
NM_012253.3 Intron NP_036385.3

View Full Product Details