Product Details
- SNP ID
-
rs9478848
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:149825080 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTCACCCAGGTACTTGACATCTGGA[G/T]TATTCTTAGCTATCTATTGTTTAAA
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
LRP11
PubMed Links
Gene Details
- Gene
- LRP11
- Gene Name
- LDL receptor related protein 11
View Full Product Details