Product Details
- SNP ID
-
rs6104066
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:45221465 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCAAGCAGCTGTGATGGGACAAAAA[A/G]GTGAGTGGAGAGGGTAAGCCTTGGG
- Phenotype
-
MIM: 182141
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SEMG2
PubMed Links
Gene Details
- Gene
- SEMG2
- Gene Name
- semenogelin II
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003008.2 |
97 |
Missense Mutation |
AGT,GGT |
S26G |
NP_002999.1 |
View Full Product Details