Product Details

SNP ID
rs6060356
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:35303415 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCATTACTGCGTCCACCATCTCA[A/G]GCAAGTTTCTAAGTCCCTGGAATAA
Phenotype
MIM: 611797
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UQCC1 PubMed Links

Gene Details

Gene
UQCC1
Gene Name
ubiquinol-cytochrome c reductase complex assembly factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184977.1 1508 UTR 3 NP_001171906.1
NM_018244.4 1508 UTR 3 NP_060714.3
NM_199487.2 1508 UTR 3 NP_955781.2
XM_011528877.1 1508 UTR 3 XP_011527179.1
XM_011528878.1 1508 UTR 3 XP_011527180.1
XM_011528879.1 1508 UTR 3 XP_011527181.1
XM_011528880.1 1508 UTR 3 XP_011527182.1
XM_011528881.2 1508 UTR 3 XP_011527183.1
XM_011528882.1 1508 UTR 3 XP_011527184.1
XM_011528883.1 1508 UTR 3 XP_011527185.1
XM_011528884.1 1508 UTR 3 XP_011527186.1

View Full Product Details