Product Details

SNP ID
rs10404390
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:713518 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCAGTGGGGGCATCTCTGCCTCT[A/G]CCCATTAGCCAGGTTCATCCTTAAG
Phenotype
MIM: 608134
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PALM PubMed Links

Gene Details

Gene
PALM
Gene Name
paralemmin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040134.1 Intron NP_001035224.1
NM_002579.2 Intron NP_002570.2
XM_005259565.4 Intron XP_005259622.1
XM_005259566.4 Intron XP_005259623.1
XM_017026850.1 Intron XP_016882339.1

View Full Product Details