Product Details

SNP ID
rs9898353
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:30124847 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCCTTACCTATAAAAAAATCAGTC[C/T]GGTTAATTCCAGCTTGTCTTTTTAC
Phenotype
MIM: 616173
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR3184 PubMed Links

Gene Details

Gene
MIR3184
Gene Name
microRNA 3184
There are no transcripts associated with this gene.

Gene
MIR423
Gene Name
microRNA 423
There are no transcripts associated with this gene.

Gene
NSRP1
Gene Name
nuclear speckle splicing regulatory protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001261467.1 Intron NP_001248396.1
NM_032141.3 Intron NP_115517.1
XM_011525345.2 Intron XP_011523647.1
XM_017025209.1 Intron XP_016880698.1
XM_017025210.1 Intron XP_016880699.1
XM_017025211.1 Intron XP_016880700.1
XM_017025212.1 Intron XP_016880701.1
XM_017025213.1 Intron XP_016880702.1
XM_017025214.1 Intron XP_016880703.1
XM_017025215.1 Intron XP_016880704.1

View Full Product Details