Product Details

SNP ID
rs10410770
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:32411498 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCCAATCTCTAGAAAGGTTGGACT[A/G]TAGAGTGAAATAACTTTTGTGTTTA
Phenotype
MIM: 613894
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DPY19L3 PubMed Links

Gene Details

Gene
DPY19L3
Gene Name
dpy-19 like 3 (C. elegans)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172774.1 Intron NP_001166245.1
NM_207325.2 Intron NP_997208.2
XM_006723045.1 Intron XP_006723108.1
XM_011526526.2 Intron XP_011524828.1
XM_017026366.1 Intron XP_016881855.1
XM_017026367.1 Intron XP_016881856.1
Gene
LOC400684
Gene Name
uncharacterized LOC400684
There are no transcripts associated with this gene.

View Full Product Details