Product Details

SNP ID
rs9436660
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:62438092 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGAAATAGCAAGGAAGATACTGCA[C/T]CTGGCAAAATGTTACATTCTCGGTG
Phenotype
MIM: 603478
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
USP1 PubMed Links

Gene Details

Gene
USP1
Gene Name
ubiquitin specific peptidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017415.1 Intron NP_001017415.1
NM_001017416.1 Intron NP_001017416.1
NM_003368.4 Intron NP_003359.3

View Full Product Details