Product Details
- SNP ID
-
rs9612413
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:23741686 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTACCATGATGAGTTCATGTTAGCC[C/T]AGCCATAAATGTTTAGGAAACTTCA
- Phenotype
-
MIM: 194544
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF70
PubMed Links
Gene Details
- Gene
- ZNF70
- Gene Name
- zinc finger protein 70
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_021916.3 |
3916 |
UTR 3 |
|
|
NP_068735.1 |
View Full Product Details