Product Details

SNP ID
rs9840783
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:125226285 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTCTTTTCTCTTTACGGTTATAT[C/T]TATGCACCATGATTTTTGCATATTT
Phenotype
MIM: 601897
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF148 PubMed Links

Gene Details

Gene
ZNF148
Gene Name
zinc finger protein 148
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021964.2 8927 UTR 3 NP_068799.2

View Full Product Details