Product Details

SNP ID
rs9324141
Assay Type
Functionally Tested
NCBI dbSNP Submissions
42
Location
Chr.1:84819690 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTGGGAAAATGTTTGAGCACATA[C/T]GCAAAAATAAATATACAAAGAGTGG
Phenotype
MIM: 605106
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
LPAR3 PubMed Links

Gene Details

Gene
LPAR3
Gene Name
lysophosphatidic acid receptor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012152.2 Intron NP_036284.1

View Full Product Details