Product Details
- SNP ID
-
rs7145500
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:65417494 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGATACTACACATTTTCCTTTTAAC[A/G]GAGGAAGAGCAAATGGGACCTATGG
- Phenotype
-
MIM: 602589
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FUT8
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76713816] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FUT8
- Gene Name
- fucosyltransferase 8
- Gene
- FUT8-AS1
- Gene Name
- FUT8 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details