Product Details

SNP ID
rs10401403
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57907154 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCTGAGCCAAAACACCCAGCTGA[A/G]ATTCTTCATATAGCTGATTATCTTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF417 PubMed Links

Gene Details

Gene
ZNF417
Gene Name
zinc finger protein 417
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297734.1 3916 Intron NP_001284663.1
NM_152475.2 3916 Intron NP_689688.2
XM_011526473.2 3916 UTR 3 XP_011524775.1
XM_017026315.1 3916 Intron XP_016881804.1

View Full Product Details