Product Details

SNP ID
rs6130901
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45712832 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTTTACTAAGTAGAAACAATCTC[C/T]TAAAACAGAAAACACCAAGTCCACA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR3617 PubMed Links

Gene Details

Gene
MIR3617
Gene Name
microRNA 3617
There are no transcripts associated with this gene.

Gene
SPINT4
Gene Name
serine peptidase inhibitor, Kunitz type 4
There are no transcripts associated with this gene.

Gene
WFDC10B
Gene Name
WAP four-disulfide core domain 10B
There are no transcripts associated with this gene.

Gene
WFDC13
Gene Name
WAP four-disulfide core domain 13
There are no transcripts associated with this gene.

Gene
WFDC3
Gene Name
WAP four-disulfide core domain 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080614.1 Intron NP_542181.1

View Full Product Details