Product Details

SNP ID
rs9875073
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:179248108 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCAGAACCCCCATTCCCCTCATT[C/T]CAGCAAACATCATTTAGGTATTAAG
Phenotype
MIM: 605222 MIM: 171834
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
KCNMB3 PubMed Links
Additional Information
For this assay, SNP(s) [rs77482668] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
KCNMB3
Gene Name
potassium calcium-activated channel subfamily M regulatory beta subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163677.1 Intron NP_001157149.1
NM_014407.3 Intron NP_055222.3
NM_171828.2 Intron NP_741979.1
NM_171829.2 Intron NP_741980.1
NM_171830.1 Intron NP_741981.1
Gene
PIK3CA
Gene Name
phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit alpha
There are no transcripts associated with this gene.

View Full Product Details