Product Details

SNP ID
rs6070734
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:59152377 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTTGGTTGAGGAGGCAGCGAGTA[A/G]GTGGACAGGTGGATTTTGGGTTTGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF831 PubMed Links

Gene Details

Gene
ZNF831
Gene Name
zinc finger protein 831
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178457.2 Intron NP_848552.1

View Full Product Details