Product Details
- SNP ID
-
rs7836454
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:30161611 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTACCTGCTCCCTTTCTCTTCATGA[C/T]TTTTGTGCTTCTTCTCATTCAGTGA
- Phenotype
-
MIM: 612963
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DCTN6
PubMed Links
Gene Details
- Gene
- DCTN6
- Gene Name
- dynactin subunit 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006571.3 |
|
Intron |
|
|
NP_006562.1 |
View Full Product Details