Product Details
- SNP ID
-
rs10172787
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:121764240 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTAGGCTGTACCACTTGTCTGGCTT[C/G]TTGGAGACACCTGACCAGTAAACAG
- Phenotype
-
MIM: 600575
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TSN
PubMed Links
Gene Details
- Gene
- TSN
- Gene Name
- translin
View Full Product Details