Product Details

SNP ID
rs10420375
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:3509655 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCGCCACCACCTGTCTCCCCTGCCT[A/C]TGTGGCTGGGCCTGTCCCGGACATT
Phenotype
MIM: 611262 MIM: 603619
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DOHH PubMed Links

Gene Details

Gene
DOHH
Gene Name
deoxyhypusine hydroxylase/monooxygenase
There are no transcripts associated with this gene.

Gene
FZR1
Gene Name
fizzy/cell division cycle 20 related 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136197.1 Intron NP_001129669.1
NM_001136198.1 Intron NP_001129670.1
NM_016263.3 Intron NP_057347.2
XM_005259573.4 Intron XP_005259630.1
XM_017026863.1 Intron XP_016882352.1

View Full Product Details