Product Details

SNP ID
rs9931183
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:982776 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCTTAGAAAGGGCTCAGCTCCTCT[A/G]GCTCCTGGACCACCCATGGGGAGGG
Phenotype
MIM: 611761 MIM: 605923
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LMF1 PubMed Links

Gene Details

Gene
LMF1
Gene Name
lipase maturation factor 1
There are no transcripts associated with this gene.

Gene
SOX8
Gene Name
SRY-box 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014587.4 Intron NP_055402.2

View Full Product Details