Product Details
- SNP ID
-
rs10247493
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:143909141 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTGAAGACTACAGGCCAATTATTTT[A/C]TATTTCCTTGTTTTGGACTTTCCTG
- Phenotype
-
MIM: 608497
MIM: 616251
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
OR2F1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78675223] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR2F1
- Gene Name
- olfactory receptor family 2 subfamily F member 1 (gene/pseudogene)
- Gene
- TCAF1
- Gene Name
- TRPM8 channel associated factor 1
There are no transcripts associated with this gene.
View Full Product Details