Product Details

SNP ID
rs10087388
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:42849782 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTATTTCAACTTAGAGTTCCCTGA[A/T]ATTGTTTTCTCATTCATTGATTCAA
Phenotype
MIM: 614649 MIM: 609520
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RNF170 PubMed Links

Gene Details

Gene
RNF170
Gene Name
ring finger protein 170
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160223.1 1859 Intron NP_001153695.1
NM_001160224.1 1859 UTR 3 NP_001153696.1
NM_001160225.1 1859 Intron NP_001153697.1
NM_030954.3 1859 Intron NP_112216.3
XM_006716404.2 1859 Intron XP_006716467.1
XM_006716405.2 1859 Intron XP_006716468.1
XM_011544666.2 1859 Intron XP_011542968.1
XM_017013881.1 1859 Intron XP_016869370.1
XM_017013882.1 1859 Intron XP_016869371.1
Gene
THAP1
Gene Name
THAP domain containing 1
There are no transcripts associated with this gene.

View Full Product Details