Product Details

SNP ID
rs10521788
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:138642843 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACTAAAGAAATGCAGTGCTTAAAT[A/G]AAAGGGCTTCATGAAATACCATGCC
Phenotype
MIM: 300070
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FGF13 PubMed Links

Gene Details

Gene
FGF13
Gene Name
fibroblast growth factor 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001139498.1 Intron NP_001132970.1
NM_001139500.1 Intron NP_001132972.1
NM_001139501.1 Intron NP_001132973.1
NM_001139502.1 Intron NP_001132974.1
NM_004114.3 Intron NP_004105.1
NM_033642.2 Intron NP_378668.1
XM_005262399.1 Intron XP_005262456.1

View Full Product Details