Product Details

SNP ID
rs6120515
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:34094354 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCAGGAAAAGATACAAAGGTGGC[C/T]TTAATTATAACTATAAAACCACCTT
Phenotype
MIM: 603908 MIM: 614663
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EIF2S2 PubMed Links

Gene Details

Gene
EIF2S2
Gene Name
eukaryotic translation initiation factor 2 subunit beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316363.1 Intron NP_001303292.1
NM_001316364.1 Intron NP_001303293.1
NM_003908.4 Intron NP_003899.2
XM_017028118.1 Intron XP_016883607.1
Gene
RALY
Gene Name
RALY heterogeneous nuclear ribonucleoprotein
There are no transcripts associated with this gene.

View Full Product Details