Product Details
- SNP ID
-
rs6124766
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:37296654 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTGCTGCCATGAACTTTTTTGTAC[C/T]TGAAGGCTTTTTCTTCTTTTGAATT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MANBAL
PubMed Links
Gene Details
- Gene
- MANBAL
- Gene Name
- mannosidase beta like
View Full Product Details