Product Details

SNP ID
rs9911864
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:64470933 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAATGCTGGCGTCCACTAGTGCCTC[A/G]TTTCTTTCTAAACCTCATTCCACAT
Phenotype
MIM: 604983
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MILR1 PubMed Links

Gene Details

Gene
MILR1
Gene Name
mast cell immunoglobulin like receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085423.1 Intron NP_001078892.1
NM_001291316.1 Intron NP_001278245.1
NM_001291317.1 Intron NP_001278246.1
XM_017024486.1 Intron XP_016879975.1
XM_017024487.1 Intron XP_016879976.1
Gene
POLG2
Gene Name
DNA polymerase gamma 2, accessory subunit
There are no transcripts associated with this gene.

View Full Product Details