Product Details
- SNP ID
-
rs9480130
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:155396950 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGCTTTAGGTCCACAGAAGAACAC[A/G]CCAATACTGCTGCTGCAGTAGGGGT
- Phenotype
-
MIM: 607105
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NOX3
PubMed Links
Gene Details
- Gene
- NOX3
- Gene Name
- NADPH oxidase 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015718.2 |
1696 |
Silent Mutation |
GGC,GGT |
G531G |
NP_056533.1 |
View Full Product Details