Product Details

SNP ID
rs5985880
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:28602541 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTATTGGAACAGACTGTGCATCA[C/G]ACGCTATTTAAACCTCATCTATATG
Phenotype
MIM: 300206
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IL1RAPL1 PubMed Links

Gene Details

Gene
IL1RAPL1
Gene Name
interleukin 1 receptor accessory protein like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014271.3 Intron NP_055086.1

View Full Product Details