Product Details

SNP ID
rs10426580
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57244853 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTTGTCTGTTTCTCTGGCCTCT[A/G]TGGGGCGTGTTTGAGGCAGCAAATG
Phenotype
MIM: 603495
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AURKC PubMed Links

Gene Details

Gene
AURKC
Gene Name
aurora kinase C
There are no transcripts associated with this gene.

Gene
ZNF805
Gene Name
zinc finger protein 805
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001023563.3 Intron NP_001018857.2
NM_001145078.1 Intron NP_001138550.1

View Full Product Details