Product Details

SNP ID
rs8192501
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:119367739 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCATGCCCTGTCCCCTTTCAGCT[A/G]TTTCCAGCATACTGTGCCCCGTCTG
Phenotype
MIM: 125950
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C2orf76 PubMed Links

Gene Details

Gene
C2orf76
Gene Name
chromosome 2 open reading frame 76
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017927.3 65 Intron NP_001017927.2
NM_001322329.1 65 Intron NP_001309258.1
NM_001322330.1 65 Intron NP_001309259.1
NM_001322331.1 65 Intron NP_001309260.1
NM_001322332.1 65 Intron NP_001309261.1
XM_017003354.1 65 Intron XP_016858843.1
Gene
DBI
Gene Name
diazepam binding inhibitor, acyl-CoA binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079862.2 65 Intron NP_001073331.1
NM_001079863.1 65 UTR 5 NP_001073332.1
NM_001178017.1 65 Intron NP_001171488.1
NM_001178041.2 65 Intron NP_001171512.1
NM_001178042.2 65 Intron NP_001171513.1
NM_001178043.2 65 Intron NP_001171514.1
NM_001282633.1 65 Intron NP_001269562.1
NM_001282634.1 65 Intron NP_001269563.1
NM_001282635.1 65 Intron NP_001269564.1
NM_001282636.1 65 Intron NP_001269565.1
NM_020548.7 65 Intron NP_065438.1

View Full Product Details