Product Details

SNP ID
rs9310985
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:32107837 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTCCCTATTGCTGGCCTGCATCA[C/T]TGCATTATAAGGAGTTATTTGATTA
Phenotype
MIM: 611778
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPD1L PubMed Links

Gene Details

Gene
GPD1L
Gene Name
glycerol-3-phosphate dehydrogenase 1-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015141.3 Intron NP_055956.1
XM_006713068.2 Intron XP_006713131.1

View Full Product Details