Product Details

SNP ID
rs6699434
Assay Type
Functionally tested
NCBI dbSNP Submissions
21
Location
Chr.1:155133540 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAGCATGAAGACCGCTGCTTGAG[A/G]TTGAAGGTGACTGTCAGTGGCAAAA
Phenotype
MIM: 605951 MIM: 191164 MIM: 613683
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DPM3 PubMed Links

Gene Details

Gene
DPM3
Gene Name
dolichyl-phosphate mannosyltransferase subunit 3
There are no transcripts associated with this gene.

Gene
EFNA1
Gene Name
ephrin A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004428.2 531 Silent Mutation AGA,AGG R142R NP_004419.2
NM_182685.1 531 Intron NP_872626.1
Gene
SLC50A1
Gene Name
solute carrier family 50 member 1
There are no transcripts associated with this gene.

View Full Product Details