Product Details
- SNP ID
-
rs10413401
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:58331215 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAACACCCAACAATTATCAAATGCA[C/T]TTTTTTTTTTTTTTTTTTTGAGACA
- Phenotype
-
MIM: 165260
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MIR6806
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs58617733] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MIR6806
- Gene Name
- microRNA 6806
There are no transcripts associated with this gene.
- Gene
- ZSCAN22
- Gene Name
- zinc finger and SCAN domain containing 22
View Full Product Details