Product Details
- SNP ID
-
rs7551203
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
17
- Location
-
Chr.1:196657973 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AACTAACAGTTAAAAGATATATTGA[C/T]ATTGAAAATAAGTAATTAGTTCTAT
- Phenotype
-
MIM: 134370
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CFH
PubMed Links
Gene Details
- Gene
- CFH
- Gene Name
- complement factor H
View Full Product Details