Product Details

SNP ID
rs9919135
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:89998226 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTATCAATGTAAAAGGCCAGTTCTA[C/T]CTATTTCAGTCCTTGTTTTGGTTCT
Phenotype
MIM: 614601
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GEMIN8P4 PubMed Links

Gene Details

Gene
GEMIN8P4
Gene Name
gem nuclear organelle associated protein 8 pseudogene 4
There are no transcripts associated with this gene.

Gene
ZNF326
Gene Name
zinc finger protein 326
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320185.1 Intron NP_001307114.1
NM_181781.3 Intron NP_861446.2
NM_182975.3 Intron NP_892020.1
NM_182976.3 Intron NP_892021.1
XM_005270779.4 Intron XP_005270836.1
XM_011541290.2 Intron XP_011539592.1

View Full Product Details