Product Details

SNP ID
hCV30063181
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:206123929 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCATAATTTAAGGATCTCTTTAT[A/G]TTCGTCACAAAGGTTATCAATGCTT
Phenotype
MIM: 157655
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GCSHP3 PubMed Links

Gene Details

Gene
GCSHP3
Gene Name
glycine cleavage system protein H pseudogene 3
There are no transcripts associated with this gene.

Gene
NDUFS1
Gene Name
NADH:ubiquinone oxidoreductase core subunit S1
There are no transcripts associated with this gene.

View Full Product Details