Product Details

SNP ID
rs6035013
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:18293411 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCAGGCCTCACTCTCTCCAGGTAG[C/T]GTAAGGGCTTCCAGCAGGATTGAAC
Phenotype
MIM: 604075
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF133 PubMed Links

Gene Details

Gene
ZNF133
Gene Name
zinc finger protein 133
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083330.2 3594 Intron NP_001076799.1
NM_001282995.1 3594 Intron NP_001269924.1
NM_001282996.1 3594 Intron NP_001269925.1
NM_001282997.1 3594 Intron NP_001269926.1
NM_001282998.1 3594 Intron NP_001269927.1
NM_001282999.1 3594 Intron NP_001269928.1
NM_001283000.1 3594 Intron NP_001269929.1
NM_001283001.1 3594 Intron NP_001269930.1
NM_001283002.1 3594 Intron NP_001269931.1
NM_001283003.1 3594 Intron NP_001269932.1
NM_001283004.1 3594 Intron NP_001269933.1
NM_001283005.1 3594 Intron NP_001269934.1
NM_001283006.1 3594 Intron NP_001269935.1
NM_001283007.1 3594 Intron NP_001269936.1
NM_001283008.1 3594 Intron NP_001269937.1
NM_003434.5 3594 Intron NP_003425.2
XM_005260819.1 3594 Intron XP_005260876.1
XM_005260820.4 3594 Intron XP_005260877.1
XM_011529336.1 3594 Intron XP_011527638.1
XM_011529337.1 3594 Intron XP_011527639.1
XM_011529338.2 3594 Intron XP_011527640.1
XM_011529339.1 3594 Intron XP_011527641.1
XM_011529340.1 3594 Intron XP_011527642.1
XM_017028041.1 3594 UTR 5 XP_016883530.1
XM_017028042.1 3594 Intron XP_016883531.1
XM_017028043.1 3594 UTR 5 XP_016883532.1
XM_017028044.1 3594 UTR 5 XP_016883533.1
XM_017028045.1 3594 UTR 5 XP_016883534.1
XM_017028046.1 3594 UTR 5 XP_016883535.1
XM_017028047.1 3594 Intron XP_016883536.1
XM_017028048.1 3594 Intron XP_016883537.1
XM_017028049.1 3594 UTR 5 XP_016883538.1
XM_017028050.1 3594 Intron XP_016883539.1
XM_017028051.1 3594 Intron XP_016883540.1
XM_017028052.1 3594 Intron XP_016883541.1
XM_017028053.1 3594 Intron XP_016883542.1
XM_017028054.1 3594 Intron XP_016883543.1
XM_017028055.1 3594 Intron XP_016883544.1
XM_017028056.1 3594 Intron XP_016883545.1
XM_017028057.1 3594 Intron XP_016883546.1
XM_017028058.1 3594 Intron XP_016883547.1

View Full Product Details