Product Details

SNP ID
rs6851081
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:122615470 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGAAACCGGGAGGTGGATCTTGCG[A/G]TGAGCAGAGATCACGCCACTGCACT
Phenotype
MIM: 605384
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IL21 PubMed Links

Gene Details

Gene
IL21
Gene Name
interleukin 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001207006.2 Intron NP_001193935.1
NM_021803.3 Intron NP_068575.1
Gene
IL21-AS1
Gene Name
IL21 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details