Product Details
- SNP ID
-
rs6685336
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
4
- Location
-
Chr.1:35818706 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGAAGGAAGGAAGGAAGGAAGGAAA[C/G]AAACAAACAGCAGATAATTCTGGAT
- Phenotype
-
MIM: 607356
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
AGO4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs28449969] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AGO4
- Gene Name
- argonaute 4, RISC catalytic component
View Full Product Details