Product Details
- SNP ID
-
rs4807209
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:2173081 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCATTGTCTAATTCCAGAACGTTCT[A/C]ATCACCCCAAAAGGAAACGCTGCCG
- Phenotype
-
MIM: 607375
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
DOT1L
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78591932] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DOT1L
- Gene Name
- DOT1 like histone lysine methyltransferase
View Full Product Details