Product Details

SNP ID
rs7516013
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:173804818 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATTTTAAGTTACATGACAAACATG[A/C]CTTCCGGATACTCAAAGAACTCTGT
Phenotype
MIM: 611503
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CENPL PubMed Links

Gene Details

Gene
CENPL
Gene Name
centromere protein L
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127181.2 Intron NP_001120653.1
NM_001171182.1 Intron NP_001164653.1
NM_033319.3 Intron NP_201576.1

View Full Product Details