Product Details

SNP ID
rs10470713
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:197749577 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAAAGGCTCGTGTGTACCGAAGTA[C/T]AGGGGACCTGTCTGCAGCAGGAGGG
Phenotype
MIM: 616933 MIM: 613516
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FYTTD1 PubMed Links

Gene Details

Gene
FYTTD1
Gene Name
forty-two-three domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001011537.2 25 UTR 5 NP_001011537.2
NM_032288.6 25 Intron NP_115664.2
Gene
RUBCN
Gene Name
RUN and cysteine rich domain containing beclin 1 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145642.3 25 UTR 5 NP_001139114.1
NM_014687.2 25 Intron NP_055502.1
XM_005269374.3 25 Intron XP_005269431.1
XM_006713827.3 25 Intron XP_006713890.1
XM_006713828.3 25 Intron XP_006713891.1
XM_006713829.3 25 UTR 5 XP_006713892.1
XM_006713830.3 25 Intron XP_006713893.1
XM_006713831.3 25 UTR 5 XP_006713894.1
XM_017007543.1 25 Intron XP_016863032.1
XM_017007544.1 25 Intron XP_016863033.1
XM_017007545.1 25 UTR 5 XP_016863034.1

View Full Product Details