Product Details
- SNP ID
-
rs5930974
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:136653876 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGATAGCAAAATTAAAAATGTCTGA[C/G]GGTCTTTTCATAGTCTGGCATTTCT
- Phenotype
-
MIM: 300386
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CD40LG
PubMed Links
Gene Details
- Gene
- CD40LG
- Gene Name
- CD40 ligand
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000074.2 |
|
Intron |
|
|
NP_000065.1 |
View Full Product Details