Product Details
- SNP ID
-
rs4241254
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:73241121 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTAAGTGTATAATTCATGGCATTA[A/G]CCAGTCTATCTTTTAAAAATAAATT
- Phenotype
-
MIM: 605140
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CCT7
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs367983565] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CCT7
- Gene Name
- chaperonin containing TCP1 subunit 7
- Gene
- PRADC1
- Gene Name
- protease associated domain containing 1
There are no transcripts associated with this gene.
View Full Product Details