Product Details

SNP ID
rs6120127
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:33161919 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGTGGTAAGCCAAGACCCCACAGA[A/C]ACAAGTCCTTCGAGGGTCCCGGACG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
BPIFA2 PubMed Links

Gene Details

Gene
BPIFA2
Gene Name
BPI fold containing family A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319164.1 Intron NP_001306093.1
NM_080574.3 Intron NP_542141.1
Gene
LOC105372714
Gene Name
uncharacterized LOC105372714
There are no transcripts associated with this gene.

View Full Product Details