Product Details

SNP ID
rs9568206
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:49450718 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATATATTTTAAATCTCCTAACATTT[C/G]CATGGTTTGGTGTGGAAGTGGCAGA
Phenotype
MIM: 612175 MIM: 607865
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CAB39L PubMed Links

Gene Details

Gene
CAB39L
Gene Name
calcium binding protein 39 like
There are no transcripts associated with this gene.

Gene
SETDB2
Gene Name
SET domain bifurcated 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160308.2 Intron NP_001153780.1
NM_001320699.1 Intron NP_001307628.1
NM_031915.2 Intron NP_114121.2
Gene
SETDB2-PHF11
Gene Name
SETDB2-PHF11 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320727.1 Intron NP_001307656.1

View Full Product Details